Merck
Activation of CO2 by phosphinoamide hafnium complexes.
Chemical communications (Cambridge, England) (2013-02-07)
Michael J Sgro, Douglas W Stephan
ABSTRACT
Hf-phosphinoamide cation complexes behave as metal-based frustrated Lewis pairs and bind one or two equivalent of CO2 and in as well can activate CO2 in a bimetallic fashion to give a pseudo-tetrahedral P2CO2 fragment linking two Hf centres.
Sigma-Aldrich
Hafnium, turnings, crystal bar, 99.7% trace metals basis