Merck
  • Molecular cytotoxic mechanisms of chlorpromazine in isolated rat hepatocytes.
Molecular cytotoxic mechanisms of chlorpromazine in isolated rat hepatocytes.
Canadian journal of physiology and pharmacology (2013-02-02)
Stephanie L MacAllister, Cheryl Young, Anna Guzdek, Nickholas Zhidkov, Peter J O'Brien
ABSTRACT
Chlorpromazine (CPZ), a member of the largest class of first-generation antipsychotic agents, is known to cause hepatotoxicity in the form of cholestasis and hepatocellular necrosis in some patients. The mechanism of CPZ hepatotoxicity is unclear, but is thought to result from reactive metabolite formation. The goal of this research was to assess potential cytotoxic mechanisms of CPZ using the accelerated cytotoxicity mechanism screening (ACMS) technique with freshly isolated rat hepatocytes. This study identified CPZ cytotoxicity and inhibition of mitochondrial membrane potential (MMP) to be concentration-dependent. Furthermore, inhibition of cytochrome P450s (CYPs), including CYP2D1 and 1A2, delayed CPZ cytotoxicity, suggesting a role for CYP activation of CPZ to a toxic metabolite(s) in this model. Metabolism studies also demonstrated glucuronide and glutathione (GSH) requirement for CPZ detoxification in hepatocytes. Inactivating the 2-electron reduction pathway, NAD(P)H quinone oxidoreductase (NQO1), caused a significant increase in hepatocyte susceptibility to CPZ, indicating quinoneimine contribution to CPZ cytotoxicity. Nontoxic concentrations of peroxidase/H(2)O(2) (inflammatory model) increased cytotoxicity in CPZ-treated hepatocytes and caused additional mitochondrial toxicity. Inflammation further depleted GSH and increased oxidized glutathione (GSSG) levels. Results suggest activation of CPZ to reactive metabolites by 2 pathways in hepatocytes: (i) a CYP-catalyzed quinoneimine pathway, and (ii) a peroxidase-catalyzed oxidation of CPZ to CPZ radicals.
Product Description
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorpromazine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)