Merck
  • High-grade prostate cancer and biochemical recurrence after radical prostatectomy among men using 5α-reductase inhibitors and alpha-blockers.
High-grade prostate cancer and biochemical recurrence after radical prostatectomy among men using 5α-reductase inhibitors and alpha-blockers.
The Prostate (2013-01-22)
Teemu J Murtola, Paula M Kujala, Teuvo L J Tammela
ABSTRACT
Two clinical trials have shown that users of 5α-reductase inhibitors finasteride and dutasteride (5-ARIs) have reduced overall prostate cancer risk, while the proportion of high-grade tumors is increased. We studied tumor characteristics, risk of biochemical recurrence and mortality after radical prostatectomy in 5-ARI and alpha-blocker users. The study cohort consisted of 1,315 men who underwent radical prostatectomy at the Tampere University Hospital during 1995-2009. Biochemical relapse was defined as serum PSA ≥ 0.2 ng/ml after the operation. Information on mortality and medication purchases was obtained from national registries. Cox proportional regression was used to analyze hazard ratios (HRs) and 95% confidence intervals (95% CI) of biochemical relapse and death. The proportion of high-grade (Gleason 7-10) tumors was significantly elevated among men who had used 5-ARIs for 4 years or longer compared to the non-users (83.3% vs. 53.3%, respectively). Survival curves for biochemical relapse-free survival differed between long-term and short-term 5-ARI users, but the hazard ratio remained statistically non-significant. Risk of biochemical recurrence was elevated among alpha-blocker users (HR 1.68, 95% CI 1.37-2.06), but in sensitivity analyses this was evident only in men using alpha-blockers after prostatectomy. Mortality was not associated with medication usage. Long-term users of finasteride or dutasteride had more often high-grade prostate cancer. Our results suggest also worse progression-free survival. The association between risk of biochemical recurrence and post-operative alpha-blocker usage suggests that voiding or storage symptoms after prostatectomy may predict biochemical relapse.
Sigma-Aldrich
Finasteride, ≥98% (HPLC), powder