Merck
  • Enantioselective synthesis of substituted piperidines by addition of aryl Grignard reagents to pyridine N-oxides.
Enantioselective synthesis of substituted piperidines by addition of aryl Grignard reagents to pyridine N-oxides.
Organic letters (2012-12-22)
Munawar Hussain, Thomas Sainte-Luce Banchelin, Hans Andersson, Roger Olsson, Fredrik Almqvist
ABSTRACT
The synthesis of optically active piperidines by enantioselective addition of aryl Grignard reagents to pyridine N-oxides and lithium binolate followed by reduction is reported for the first time. The reaction results in high yields (51-94%) in combination with good ee (54-80%). Some of these products were subsequently recrystallized, affording enhanced optical purities (>99% ee).
Sigma-Aldrich
Pyridine N-oxide, 95%