Merck
  • High on/off ratio field effect transistors based on exfoliated crystalline SnS2 nano-membranes.
High on/off ratio field effect transistors based on exfoliated crystalline SnS2 nano-membranes.
Nanotechnology (2012-12-15)
Debtanu De, John Manongdo, Sean See, Vincent Zhang, Arnold Guloy, Haibing Peng
ABSTRACT
We report the implementation of field effect transistors based on exfoliated nano-membranes of a layered two-dimensional semiconductor SnS(2), which exhibit an on/off ratio exceeding 2 × 10(6) and a carrier mobility of ∼1 cm(2) V(-1) s(-1). The results demonstrate the great potential of SnS(2), a layered semiconductor with finite band gap, as the building block for future nanoelectronic applications complementary to graphene-based materials with zero or small band gaps.