Merck
  • Randomized phase III placebo-controlled trial of letrozole plus oral temsirolimus as first-line endocrine therapy in postmenopausal women with locally advanced or metastatic breast cancer.
Randomized phase III placebo-controlled trial of letrozole plus oral temsirolimus as first-line endocrine therapy in postmenopausal women with locally advanced or metastatic breast cancer.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2012-12-13)
Antonio C Wolff, Ann A Lazar, Igor Bondarenko, August M Garin, Stephen Brincat, Louis Chow, Yan Sun, Zora Neskovic-Konstantinovic, Rodrigo C Guimaraes, Pierre Fumoleau, Arlene Chan, Soulef Hachemi, Andrew Strahs, Maria Cincotta, Anna Berkenblit, Mizue Krygowski, Lih Lisa Kang, Laurence Moore, Daniel F Hayes
ABSTRACT
Recent data showed improvement in progression-free survival (PFS) when adding everolimus to exemestane in patients with advanced breast cancer experiencing recurrence/progression after nonsteroidal aromatase inhibitor (AI) therapy. Here, we report clinical outcomes of combining the mammalian target of rapamycin (mTOR) inhibitor temsirolimus with letrozole in AI-naive patients. This phase III randomized placebo-controlled study tested efficacy/safety of first-line oral letrozole 2.5 mg daily/temsirolimus 30 mg daily (5 days every 2 weeks) versus letrozole/placebo in 1,112 patients with AI-naive, hormone receptor-positive advanced disease. An independent data monitoring committee recommended study termination for futility at the second preplanned interim analysis (382 PFS events). Patients were balanced (median age, 63 years; 10% stage III, 40% had received adjuvant endocrine therapy). Those on letrozole/temsirolimus experienced more grade 3 to 4 events (37% v 24%). There was no overall improvement in primary end point PFS (median, 9 months; hazard ratio [HR], 0.90; 95% CI, 0.76 to 1.07; P = .25) nor in the 40% patient subset with prior adjuvant endocrine therapy. An exploratory analysis showed improved PFS favoring letrozole/temsirolimus in patients ≤ age 65 years (9.0 v 5.6 months; HR, 0.75; 95% CI, 0.60 to 0.93; P = .009), which was separately examined by an exploratory analysis of 5-month PFS using subpopulation treatment effect pattern plot methodology (P = .003). Adding temsirolimus to letrozole did not improve PFS as first-line therapy in patients with AI-naive advanced breast cancer. Exploratory analyses of benefit in younger postmenopausal patients require external confirmation.
Sigma-Aldrich
Letrozole, ≥98% (HPLC)
Letrozole, European Pharmacopoeia (EP) Reference Standard