- Neuroprotective effects of Cyperus rotundus on SIN-1 induced nitric oxide generation and protein nitration: ameliorative effect against apoptosis mediated neuronal cell damage.
Neuroprotective effects of Cyperus rotundus on SIN-1 induced nitric oxide generation and protein nitration: ameliorative effect against apoptosis mediated neuronal cell damage.
Nitrosylation of tyrosine (3-nitro tyrosine, 3-NT) has been implicated in the pathophysiology of various disorders particularly neurodegenerative conditions and aging. Cyperus rotundus rhizome is being used as a traditional folk medicine to alleviate a variety of disorders including neuronal stress. The herb has recently found applications in food and confectionary industries also. In current study, we have explored the protective effects of C. rotundus rhizome extract (CRE) through its oxido-nitrosative and anti apoptotic mechanism to attenuate peroxynitrite (ONOO(-)) induced neurotoxicity using human neuroblastoma SH-SY5Y cells. Our results elucidate that pre-treatment of neurons with CRE ameliorates the mitochondrial and plasma membrane damage induced by 500 μM SIN-1 to 80% and 24% as evidenced by MTT and LDH assays. CRE inhibited NO generation by downregulating i-NOS expression. SIN-1 induced depletion of antioxidant enzyme status was also replenished by CRE which was confirmed by immunoblot analysis of SOD and CAT. The CRE pre-treatment efficiently potentiated the SIN-1 induced apoptotic biomarkers such as bcl-2 and caspase-3 which orchestrate the proteolytic damage of the cell. The ONOO(-) induced damage to cellular, nuclear and mitochondrial integrity was also restored by CRE. Furthermore, CRE pre-treatment also regulated the 3-NT formation which shows the potential of plant extract against tyrosine nitration. Taken together, our findings suggest that CRE might be developed as a preventive agent against ONOO(-) induced apoptosis.