Merck
  • A molecular model of artificial photosynthesis: Mn-Ca binuclear complex for photosynthetic oxidation of water.
A molecular model of artificial photosynthesis: Mn-Ca binuclear complex for photosynthetic oxidation of water.
Indian journal of biochemistry & biophysics (2012-10-20)
Nibedita Mishra, Pranab Kishor Mohapatra, Mukesh Kumar Raval, Basanti Biswal, Uday Chand Biswal
ABSTRACT
The redox active component of oxygenic photosynthetic reaction center II contains metal cluster Mn4-Ca, where two H2O are oxidized to O2 and four H+ ions are liberated. A binuclear Mn-Ca metal center binding one substrate H2O on each ion is proposed to be the minimal unit of the redox center. A model for the water oxidizing metal cluster is built with molecular modeling software (HyperChem 8.0 Pro). Mn, being a transitional metal with variable valency is redox active, while Ca is redox inert. Formation and deprotonation of H2O+ on MnIII may be favorable compared to Ca. Deprotonation of H2O+ yields a stable species HO(-) on MnIV by transfer of one electron from MnIII as a consequence of first photoact. Similarly, during second photoact, it may lead to formation of MnV = O. The O-O bond may be formed in the third photoact between O on Mn and H2O on Ca. Subsequently, HO2*(-) may be formed, leading to formation of O2. Molecular models are built for each transition states.
Product Description
Supelco
4-Methylphenol solution, certified reference material, 5000 μg/mL in methanol
Supelco
p-Cresol, analytical standard
Sigma-Aldrich
p-Cresol, 99%
Sigma-Aldrich
p-Cresol, ≥99%, FG