Merck
  • Conductance quantization and synaptic behavior in a Ta2O5-based atomic switch.
Conductance quantization and synaptic behavior in a Ta2O5-based atomic switch.
Nanotechnology (2012-10-13)
Tohru Tsuruoka, Tsuyoshi Hasegawa, Kazuya Terabe, Masakazu Aono
ABSTRACT
Quantized conductance was observed in a cation-migration-based resistive switching memory cell with a simple metal-insulator-metal (MIM) structure using a thin Ta(2)O(5) layer. The observed conductance changes are attributed to the formation and dissolution of a metal filament with an atomic point contact of different integer multiples in the Ta(2)O(5) layer. The results demonstrate that atomic point contacts can be realized in an oxide-based MIM structure that functions as a nanogap-based atomic switch (Terabe et al 2005 Nature 433 47). By applying consecutive voltage pulses at periodic intervals of different times, we also observed an effect analogous to the long-term potentiation of biological synapses, which shows that the oxide-based atomic switch has potential for use as an essential building block of neural computing systems.
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 99.99% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis