Merck
Recurrent neonatal organophoshorus poisoning.
Indian pediatrics (2012-10-02)
Yusuf Parvez, Aji Mathew, Satheesh Kalantra Kutti
ABSTRACT
Organophosphorus poisoning in neonates is extremely rare and needs high index of suspicion to diagnose it. The clinical presentation is often confused with the features of sepsis like apnea, copious oral secretions, diarrhea, letharginess, seizures. There may be recurrence of manifestations due to chronic exposure. We report a classic case admitted in the intensive care unit of our hospital.
Product Description
Supelco
Diazinon solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Supelco
Diazinon, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Pyridine-2-aldoxime methochloride
Sigma-Aldrich
2-Pyridinealdoxime methiodide, 99%
Supelco
Diazinon, PESTANAL®, analytical standard