Merck
  • Captopril avoids hypertension, the increase in plasma angiotensin II but increases angiotensin 1-7 and angiotensin II-induced perfusion pressure in isolated kidney in SHR.
Captopril avoids hypertension, the increase in plasma angiotensin II but increases angiotensin 1-7 and angiotensin II-induced perfusion pressure in isolated kidney in SHR.
Autonomic & autacoid pharmacology (2012-09-22)
P Castro-Moreno, J P Pardo, R Hernández-Muñoz, J J López-Guerrero, L Del Valle-Mondragón, G Pastelín-Hernández, M Ibarra-Barajas, R Villalobos-Molina
ABSTRACT
We investigated captopril effects, an ACE inhibitor, on hypertension development, on Ang II and Ang-(1-7) plasma concentrations, on Ang II-induced contraction in isolated kidneys, and on kidney AT1R from spontaneously hypertensive (SHR) rats. Five weeks-old SHR and Wistar Kyoto (WKY) rats were treated with captopril at 30 mg/kg/day, in drinking water for 2 or 14 weeks. Systolic blood pressure (SBP) was measured, and isolated kidneys were tested for perfusion pressure and AT1R expression; while Ang II and Ang-(1-7) concentrations were determined in plasma. Captopril did not modify SBP in WKY rats and avoided its increase as SHR aged. Plasma Ang-II concentration was ∼4-5 folds higher in SHR rats, and captopril reduced it (P<0.05); while captopril increased Ang-(1-7) by ∼2 fold in all rat groups. Captopril increased Ang II-induced pressor response in kidneys of WKY and SHR rats, phenomenon not observed in kidneys stimulated with phenylephrine, a α₁-adrenoceptor agonist. Captopril did not modify AT1R in kidney cortex and medulla among rat strains and ages. Data indicate that captopril increased Ang II-induced kidney perfusion pressure but not AT₁R density in kidney of WKY and SHR rats, due to blockade of angiotensin II synthesis; however, ACE inhibitors may have other actions like activating signaling processes that could contribute to their diverse effects.
Sigma-Aldrich
Captopril, meets USP testing specifications
Supelco
Captopril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder