Merck
  • Nonsuture anastomosis of arteries and veins using the magnetic pinned-ring device: a histologic and scanning electron microscopic study.
Nonsuture anastomosis of arteries and veins using the magnetic pinned-ring device: a histologic and scanning electron microscopic study.
Annals of vascular surgery (2012-07-28)
Shi-qi Liu, Peng Lei, Zhu-ping Cao, Yi Lv, Jian-hui Li, Xiao-hai Cui
ABSTRACT
The goal of this study was to evaluate the performance of the magnetic pinned-ring device for nonsuture vascular anastomosis. The magnetic pinned-ring device consists of paired magnetic rings that are coated with titanium nitride and embedded in a polypropylene shell; the rings are equipped with alternately spaced holes and titanium pins. The vascular anastomosis procedure using the novel magnetic pinned-ring device was performed on 14 mongrel dogs, and the traditional hand-sewing technique was used on 14 additional dogs. In situ end-to-end anastomoses were performed in the femoral artery and the inferior vena cava. Patency was confirmed through ultrasonographic scans at different time points as late as 24 weeks after surgery. Gross observation, histological staining, and scanning electron microscopy were used to evaluate the results at 24 weeks postoperatively. The time required to perform the vascular anastomosis was significantly shorter for the magnetic device than for hand sewing. A continuity of re-endothelialization was confirmed in all anastomotic stomas after 24 weeks, and neither formation of aneurysms nor thickening of the vascular wall was noted. The re-endothelialization was smooth at the anastomotic site of the magnetic device, whereas hand sewing resulted in rough and uneven re-endothelialization and the presence of visible sutures. Moreover, the endothelial cells were regularly arranged at the anastomotic site of the magnetic device, whereas different-sized and irregularly aligned endothelial cells were present at the hand-sewn anastomotic site. Use of the magnetic device was associated with significantly decreased deposition of fibrotic collagen and depressed infiltration of inflammatory cells compared with use of the hand-sewing technique. The magnetic pinned-ring device offers a simple, fast, reliable, and efficacious technique for nonsuture vascular anastomosis. Use of this device shortens operation time, maintains a high patency rate, and improves the healing of vascular tissue.
Sigma-Aldrich
Titanium nitride, <3 μm