Merck
  • Analysis of the role of Nrf2 in the expression of liver proteins in mice using two-dimensional gel-based proteomics.
Analysis of the role of Nrf2 in the expression of liver proteins in mice using two-dimensional gel-based proteomics.
Pharmacological reports : PR (2012-07-21)
Azman Abdullah, Neil R Kitteringham, Rosalind E Jenkins, Christopher Goldring, Larry Higgins, Masayuki Yamamoto, John Hayes, B Kevin Park
ABSTRACT
The transcription factor Nrf2 regulates expression of multiple cellular defence proteins through the antioxidant response element (ARE). Nrf2-deficient mice (Nrf2(-/-)) are highly susceptible to xenobiotic-mediated toxicity, but it is not known whether this reflects low basal expression or reduced inducibility of Nrf2-regulated genes in response to chemical insults. Wild type and Nrf2(-/-) mice were fed diet supplemented with the established Nrf2 inducer butylated hydroxyanisole (BHA) [0.5% (w/w)] for 14 days. To define the range of Nrf2-regulated proteins, both basally and following exposure to BHA, a comparison of the liver proteomes of Nrf2(-/-) and wild type mice was conducted. The two-dimensional gel electrophoresis (2-DE) technique and MALDI mass spectrometry were utilized in the attempt to define Nrf2-regulated proteins. Overall, 24 proteins were identified, which were regulated either basally (3 proteins), inducibly (16 proteins), or both (5 proteins). These included several well-established Nrf2-driven gene products e.g., aldo-keto reductase and glutathione transferases. Multiple consensus ARE/ARE-like sequences were found in the Nrf2-regulated genes. This study confirms the central role of Nrf2 in the induction of multiple defense proteins as well as its control in the constitutive expression of certain proteins.
Sigma-Aldrich
Butylated hydroxyanisole, ≥98.5%
Sigma-Aldrich
Butylated hydroxyanisole, 99%, FCC, FG
Butylhydroxyanisole, European Pharmacopoeia (EP) Reference Standard