Merck
  • Involvement of different types of potassium channels in the antidepressant-like effect of ascorbic acid in the mouse tail suspension test.
Involvement of different types of potassium channels in the antidepressant-like effect of ascorbic acid in the mouse tail suspension test.
European journal of pharmacology (2012-05-12)
Morgana Moretti, Josiane Budni, Camille Mertins Ribeiro, Ana Lúcia S Rodrigues
ABSTRACT
Considering that the administration of ascorbic acid elicits an antidepressant-effect in mice by a mechanism which involves an interaction with N-methyl-D-aspartate receptors and the l-arginine-nitric oxide-cGMP pathway and taking into account that the stimulation of this pathway is associated with the activation of potassium (K⁺) channels, this study investigated the involvement of different types of K⁺ channels on the effect of ascorbic acid in the mouse tail suspension test (TST). Intracerebroventricular administration of tetraethylammonium (TEA, a non-specific blocker of K⁺ channels, 25 pg/site), glibenclamide (an ATP-sensitive K⁺ channel blocker, 0.5 pg/site), charybdotoxin (a large- and intermediate conductance calcium-activated K⁺ channel blocker, 25 pg/site) or apamin (a small-conductance calcium-activated K⁺ channel blocker, 10 pg/site) was able to produce a synergistic effect with a sub-effective dose of ascorbic acid (0.1 mg/kg) given orally (p.o.). The antidepressant-like effect of ascorbic acid (1 mg/kg, p.o.) in the TST was prevented by the pre-treatment of mice with cromakalim (a K⁺ channel opener, 10 μg/site, i.c.v.) and minoxidil (10 μg/site, i.c.v.). Moreover, cromakalim abolished the synergistic effect elicited by the combined treatment with sub-effective doses of ascorbic acid and 7-nitroindazole. The administration of the K⁺ channel modulators alone or in combination with ascorbic acid did not affect the locomotion of mice. Together, our results show that the antidepressant-like effect of ascorbic acid in the TST may involve, at least in part, the modulation of neuronal excitability, via inhibition of K⁺ channels.
Sigma-Aldrich
Apamin, synthetic, ≥97% (HPLC)
Sigma-Aldrich
Apamin, from bee venom, ≥95% (HPLC)
Sigma-Aldrich
Charybdotoxin, ≥90% (HPLC)