Merck
  • Ginsenoside Rg3 attenuates cell migration via inhibition of aquaporin 1 expression in PC-3M prostate cancer cells.
Ginsenoside Rg3 attenuates cell migration via inhibition of aquaporin 1 expression in PC-3M prostate cancer cells.
European journal of pharmacology (2012-03-20)
Xue-Yang Pan, Hao Guo, Jing Han, Feng Hao, Yu An, Yan Xu, Yilixiati Xiaokaiti, Yan Pan, Xue-Jun Li
ABSTRACT
Ginsenoside Rg3 (Rg3), one of the bioactive extracts found in ginseng root, was reported to have anti-cancer activity in various cancer models. The anti-proliferation effect of Rg3 on prostate cancer cells has been well reported. To test whether Rg3 has an anti-metastatic effect on prostate cancer, we treated a highly metastatic PC-3M prostate cancer cell line with Rg3. We found that Rg3 (10μM) led to remarkable inhibition of PC-3M cell migration. Simultaneously, exposure to Rg3 suppressed expression of the aquaporin 1 (AQP1) water channel protein, which has previously been reported to be involved in cell migration. Overexpression of AQP1 attenuated Rg3-induced inhibition of cell migration, and introduction of a shRNA targeting AQP1 abrogated the inhibitory effect of Rg3, although the basal level of cell migration was decreased by RNA interference. In mechanism study, estrogen receptor- and glucocorticoid receptor-dependent pathways are proved uninvolved in the AQP1 regulation by Rg3. However, Rg3 treatment triggered the activation of p38 MAPK; and SB202190, a specific inhibitor of p38 MAPK, antagonized the Rg3-induced regulation of AQP1 and cell migration, suggesting a crucial role for p38 in the regulation process. Deletion analysis of the promoter region of AQP1 was also conducted using dual-luciferase assay, which indicated that the -1000 bp to -200 bp promoter region was involved in the AQP1 regulation by Rg3. In all, we conclude that Rg3 effectively suppresses migration of PC-3M cells by down-regulating AQP1 expression through p38 MAPK pathway and some transcription factors acting on the AQP1 promoter.
Sigma-Aldrich
Ginsenoside Rg3, ≥98% (HPLC)
Supelco
Ginsenoside Rg3, analytical standard