Merck
  • Multicatalytic asymmetric synthesis of complex tetrahydrocarbazoles via a Diels-Alder/benzoin reaction sequence.
Multicatalytic asymmetric synthesis of complex tetrahydrocarbazoles via a Diels-Alder/benzoin reaction sequence.
Organic letters (2012-03-03)
Yankai Liu, Manuel Nappi, Eduardo C Escudero-Adán, Paolo Melchiorre
ABSTRACT
Expanding upon the recently developed aminocatalytic asymmetric indole-2,3-quinodimethane strategy, a straightforward synthesis of structurally and stereochemically complex tetrahydrocarbazoles has been devised. The chemistry's complexity-generating power was further harnessed by designing a multicatalytic, one-pot Diels-Alder/benzoin reaction sequence to stereoselectively access trans-fused tetracyclic indole-based compounds having four stereogenic centers with very high fidelity.
Sigma-Aldrich
Benzoin, purified by sublimation, ≥99.5%
Sigma-Aldrich
Benzoin, 98%