Merck
  • Iodine bonding stabilizes iodomethane in MIDAS pesticide. Theoretical study of intermolecular interactions between iodomethane and chloropicrin.
Iodine bonding stabilizes iodomethane in MIDAS pesticide. Theoretical study of intermolecular interactions between iodomethane and chloropicrin.
Journal of agricultural and food chemistry (2012-02-09)
Rainer Glaser, Kaitlan Prugger
ABSTRACT
The results are reported of a theoretical study of iodomethane (H(3)C-I, 1) and chloropicrin (Cl(3)C-NO(2), 2), of the heterodimers 3-6 formed by aggregation of 1 and 2, and of their addition products 7 and 8 and their possible fragmentation reactions to 9-18. Mixtures of iodomethane and chloropicrin are not expected to show chemistry resulting from their reactions with each other. The structures and stabilities are discussed of the iodine-bonded molecular aggregates (IBMA) 3 and 4 and of the hydrogen- and iodine-bonded molecular aggregates (IHBMA) 5 and 6. The mixed aggregates 3-5 are bound on the free enthalpy surface relative to the homodimers of 1 and 2, and the IBMA structures 3 and 4 are most stable. This result suggests that the mixture of chloropicrin and iodomethane in the pesticide Midas is a good choice to reduce the volatility of iodomethane because of thermodynamically stabilizing iodine bonding.
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)
Sigma-Aldrich
Iodomethane-13C, 99 atom % 13C, 99% (CP), contains copper as stabilizer
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99.5%
Sigma-Aldrich
Iodomethane-d3, ≥99.5 atom % D, ≥99% (CP), contains copper as stabilizer
Sigma-Aldrich
Iodomethane solution, 2.0 M in tert-butyl methyl ether, contains copper as stabilizer