Merck
  • Novel flexible dry PU/TiN-multipin electrodes: first application in EEG measurements.
Novel flexible dry PU/TiN-multipin electrodes: first application in EEG measurements.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference (2012-01-19)
P Fiedler, P Pedrosa, S Griebel, C Fonseca, F Vaz, F Zanow, J Haueisen
ABSTRACT
Dry biosignal electrodes for electro-encephalography (EEG) are an essential step for realization of ubiquitous EEG monitoring and brain computer interface technologies. We propose a novel electrode design with a specific shape for hair layer interfusion and reliable skin contact. An electrically conductive Titanium-Nitride (TiN) thin layer is deposited on a polyurethane substrate using a multiphase DC magnetron sputtering technique. In the current paper we describe the development and manufacturing of the electrode. Furthermore, we perform comparative EEG measurements with conventional Ag/AgCl electrodes in a 6-channel setup. Our results are promising, as the primary shape of the EEG is preserved in the signals of both electrodes sets, according to recordings of spontaneous EEG and visual evoked potentials. The variance of both signals is in the same order of magnitude. The Wilcoxon-Mann-Whitney two-sample rank-sum test revealed no significant differences for 25 of the 28 compared signal episodes. Hence, our novel electrodes show equivalent signal quality compared to conventional Ag/AgCl electrodes.
Sigma-Aldrich
Titanium nitride, <3 μm