Merck
The level of arabinitol in autistic children after probiotic therapy.
Nutrition (Burbank, Los Angeles County, Calif.) (2011-11-15)
Joanna Kałużna-Czaplińska, Sylwia Błaszczyk
ABSTRACT
The level of D-arabinitol (DA) and the ratio of D-/L-arabinitol (DA/LA) in the urine of children with autism were investigated. The changes in DA/LA after probiotic treatment in urine samples of children with autism were studied. DA and LA and the DA/LA ratio were identified by capillary gas chromatography/mass spectrometry in urine before and after the probiotic therapy. The level of DA is significantly higher (P < 0.05) in the urine of autistic children before (A) and after probiotic supplementation (A1) (160.04 ± 22.88 μmol/mmol creatinine and 89.53 ± 37.41 μmol/mmol creatinine, respectively). Nonetheless, the probiotic supplementation let to a significant decrease in DA and DA/LA and to a significant improvement in ability of concentration and carrying out orders. The use of probiotics seems to be helpful in reducing the level of DA and the ratio of DA/LA in the urine of children with autism.
Product Description
L-Arabinitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-(−)-Arabitol, ≥98% (GC)
Sigma-Aldrich
D-(+)-Arabitol, ≥99% (GC)