Merck
  • Determination of body water compartments in neonatal foals by use of indicator dilution techniques and multifrequency bioelectrical impedance analysis.
Determination of body water compartments in neonatal foals by use of indicator dilution techniques and multifrequency bioelectrical impedance analysis.
American journal of veterinary research (2011-10-04)
C Langdon Fielding, K Gary Magdesian, Judy E Edman
ABSTRACT
To determine values for total body water (TBW), extracellular fluid volume (ECFV), intracellular fluid volume (ICFV), and plasma volume (PV) in healthy neonatal (< 24 hours old) foals and to create a multifrequency bioelectrical impedance analysis (MF-BIA) model for use in neonatal foals. 7 healthy neonatal foals. Deuterium oxide (0.4 g/kg, IV), sodium bromide (30 mg/kg, IV), and Evans blue dye (1 mg/kg, IV) were administered to each foal. Plasma samples were obtained following an equilibration period, and the TBW, ECFV, ICFV, and PV were calculated for each foal. An MF-BIA model was created by use of morphometric measurements from each foal. Mean ± SD values were obtained for TBW (0.744 ± 0.024 L/kg), ICFV (0.381 ± 0.018 L/kg), ECFV (0.363 ± 0.014 L/kg), and PV (0.096 ± 0.015 L/kg). The 95% limits of agreement between the MF-BIA and indicator dilution techniques were within ± 2 L for TBW and ECFV. Fluid volumes in neonatal foals were found to be substantially larger than fluid volumes in adult horses. Multifrequency bioelectrical impedance analysis may be a useful technique for predicting TBW, ICFV, and ECFV in neonatal foals.
Product Description
Supelco
Density Standard 1251 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium bromide, ACS reagent, ≥99.0%