Merck
  • Application of QuEChERS method for extraction of selected persistent organic pollutants in fish tissue and analysis by gas chromatography mass spectrometry.
Application of QuEChERS method for extraction of selected persistent organic pollutants in fish tissue and analysis by gas chromatography mass spectrometry.
Journal of chromatography. A (2011-09-09)
Hans Ragnar Norli, Agnethe Christiansen, Ermias Deribe
ABSTRACT
The QuEChERS method developed for 22 organochlorine pesticides (OCPs) and 7-polychlorinated biphenyls (PCBs) in fish tissue involves a simple and efficient freezing technique for removal of lipids. The equipment developed consists of disposable syringes and a freezing block constructed from simple materials found in most laboratories. The freezing block keeps the temperature in the extract at -20.5°C up to 10 min after being exposed to room temperature. After the freezing step 69% of the lipids in tilapia and 61% in salmon are removed. Further reduction of co-extractives up to 96% in tilapia and 87% in salmon can be made by treatment with CaCl(2) and primary secondary amino sorbent (PSA) which removes the fatty acids. Spiking experiments in tilapia at 5 and 50 ng/g and extracted with acetonitrile show recovery range from 70 to 115% for all compounds. In salmon the recoveries are in the range 43-118% for the OCPs and 26-65% for the PCBs. Analysis of a standard reference material shows acceptable results for most of the pesticides but low results for the PCBs. The estimated LOQs were in the range 1-5 ng/g for tilapia and 2-10 ng/g for salmon. The method has been applied to analyse fish samples from Lake Koka in Ethiopia. It was investigated if addition of a less polar water miscible solvent than acetonitrile could increase the recovery of OCPs and PCBs. The results show that a mixture of 75% acetonitrile and 25% tetrahydrofurane (ACN/THF 75/25) clearly enhances the recoveries for most OCPs (47-101%) and PCBs (42-79%) from salmon. The recovery of aldrin increases significantly from 55% to over 80%. The method using ACN/THF 75/25 is applicable to the extraction of OCPs and PCBs from fish tissue having a lipid content of up to about 11% (salmon) with recoveries ≥70% for most of the OCPs and ≥42% for the PCBs.
Product Description
Supelco
Aldrin solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Supelco
Aldrin, PESTANAL®, analytical standard