Merck
  • Efficient and scalable one-pot synthesis of 2,4-dienols from cycloalkenones: optimized total synthesis of valerenic acid.
Efficient and scalable one-pot synthesis of 2,4-dienols from cycloalkenones: optimized total synthesis of valerenic acid.
Organic letters (2011-09-08)
Juergen Ramharter, Johann Mulzer
ABSTRACT
A mild and selective one-pot procedure to provide 2,4-dienols from simple cycloalkenones in high yields is described. This transformation is based on the in situ formation of acid-labile allylic alcohols, which on treatment with trifluoroacetic acid undergo a formal [1,3]-hydroxy migration to form diastereo- and enantiomerically enriched 2,4-dienols. The usefulness of this protocol is demonstrated in a short synthesis of valerenic acid.
Product Description
Valerenic acid, primary reference standard
Supelco
Valerenic acid, analytical standard