Merck
Synthesis of thermosensitive microgels with a tunable magnetic core.
Langmuir : the ACS journal of surfaces and colloids (2011-07-28)
Marco Laurenti, Pablo Guardia, Rafael Contreras-Cáceres, Jorge Pérez-Juste, Antonio Fernandez-Barbero, Enrique Lopez-Cabarcos, Jorge Rubio-Retama
ABSTRACT
In this work, we describe a new methodology for the preparation of monodisperse and thermosensitive microgels with magnetic core. In order to produce such a material, hydrophobic magnetic Fe(3)O(4) nanoparticles were prepared by two methods: thermal decomposition and coprecipitation. The surface of these nanoparticles was modified by addition of 3-butenoic acid, and after that these nanoparticles were dispersed in water and submitted to free radical polymerization at 70 °C in the presence of N-isopropylacrylamide (NIPAM) and bisacrylamide. The result of this reaction was monodisperse microgels with a magnetic core. By varying the amount of 3-butenoic acid, it was possible to obtain hybrid microgels with different magnetic core sizes and different architectures.
Sigma-Aldrich
3-Butenoic acid, 97%