Merck
  • Immunopotentiation and antitumor effects of a ginsenoside Rg₃-fortified red ginseng preparation in mice bearing H460 lung cancer cells.
Immunopotentiation and antitumor effects of a ginsenoside Rg₃-fortified red ginseng preparation in mice bearing H460 lung cancer cells.
Environmental toxicology and pharmacology (2011-07-27)
Dongsun Park, Dae-Kwon Bae, Jeong Hee Jeon, Jinsoo Lee, Namgil Oh, Goeun Yang, Yun-Hui Yang, Tae Kyun Kim, Jugyeong Song, Sun Hee Lee, Byeng Sub Song, Tae Hawn Jeon, Shin Jyung Kang, Seong Soo Joo, Seung U Kim, Yun-Bae Kim
ABSTRACT
Antitumor effects of a ginsenoside Rg(3)-fortified red ginseng preparation (Rg(3)-RGP) were investigated in human non-small cell lung carcinoma (H460) cells using in vitro cytotoxicity assay and in vivo nude mouse xenograft model. Immunomodulatory effects of the preparation were also assessed by measuring the facilitating activities on the nitric oxide (NO) release from peritoneal macrophages, in vitro and in vivo lymphocyte proliferation, and the carbon clearance from circulating blood. In a cell level, Rg(3)-RGP exerted H460 cytotoxicity and facilitated splenocyte proliferation at very high concentrations, without affecting NO production. However, oral administration of Rg(3)-RGP (100-300 mg/kg) enhanced carbon particle-phagocytic index of blood macrophages up to 360-397% of control value. In addition, Rg(3)-RGP significantly increased the splenocyte proliferation (23% at 100mg/kg). In tumor-bearing mice, 28-day oral treatment with Rg(3)-RGP (100mg/kg) remarkably suppressed the tumor growth, leading to the decrease of the tumor volume and weight by 30-31%, which was comparable to the effect (27-29% reduction) of doxorubicin (2mg/kg at 3-day intervals). While Rg(3)-RGP did not cause adverse effects, intravenous injection of doxorubicin markedly decreased body and testes weights, and exhibited severe depletion of spermatogenic cells in the atrophic seminiferous tubules. These results indicate that Rg(3)-RGP exerts antitumor activities via indirect immunomodulatory actions, without causing adverse effects as seen in doxorubicin.
Sigma-Aldrich
Ginsenoside Rg3, ≥98% (HPLC)
Supelco
Ginsenoside Rg3, analytical standard