Merck
  • A novel water-soluble NHC-Pd polymer: an efficient and recyclable catalyst for the Suzuki coupling of aryl chlorides in water at room temperature.
A novel water-soluble NHC-Pd polymer: an efficient and recyclable catalyst for the Suzuki coupling of aryl chlorides in water at room temperature.
Chemical communications (Cambridge, England) (2011-06-11)
Babak Karimi, Pari Fadavi Akhavan
ABSTRACT
A novel water-soluble NHC-Pd polymer with triethylene glycol legs was developed as a water soluble and highly recyclable catalyst that shows high catalytic activity in the Suzuki-Miyaura coupling of aryl chlorides in high yields in water at room temperature.
Sigma-Aldrich
Triethylene glycol, BioUltra, anhydrous, ≥99.0% (GC)
Sigma-Aldrich
Triethylene glycol, ReagentPlus®, 99%