Merck
  • 1,3-Dipolar cycloaddition-decarboxylation reactions of an azomethine ylide with isatoic anhydrides: formation of novel benzodiazepinones.
1,3-Dipolar cycloaddition-decarboxylation reactions of an azomethine ylide with isatoic anhydrides: formation of novel benzodiazepinones.
Organic letters (2010-12-24)
Asha M D'Souza, Nadia Spiccia, Jose Basutto, Pawel Jokisz, Leon S-M Wong, Adam G Meyer, Andrew B Holmes, Jonathan M White, John H Ryan
ABSTRACT
A nonstabilized azomethine ylide reacts with a wide range of substituted isatoic anhydrides to afford novel 1,3-benzodiazepin-5-one derivatives, which are generally isolated in high yield. The transformations involve 1,3-dipolar cycloaddition reactions of the ylide with the anhydrides to give transient, and in a representative case spectroscopically observable, oxazolidine intermediates that undergo ring-opening-decarboxylation-ring-closing reaction cascades to yield the 1,3-benzodiazepin-5-one products.
Sigma-Aldrich
Isatoic anhydride, 96%