Merck
  • Two-dimensional self-assemblies of silica nanoparticles formed using the "bubble deposition technique".
Two-dimensional self-assemblies of silica nanoparticles formed using the "bubble deposition technique".
Langmuir : the ACS journal of surfaces and colloids (2010-10-06)
Xinfeng Zhang, Guolei Tang, Shihe Yang, Jean-Jacques Benattar
ABSTRACT
Two-dimensional silica nanoparticle assemblies were obtained by deposition of bubble made from a surfactant solution containing nanoparticles onto hydrophobic silicon substrate. The morphologies of the nanoparticle assemblies can be finely controlled by several experimental parameters, including surfactant concentration, nanoparticle concentration, and deposition time. Monolayer of nanoparticles with surface coverage of about 100% can be obtained under appropriate conditions. The method can also be applied to another hydrophobic substrate, HMDS (hexamethyldisilazane)-modified silicon substrate. Furthermore, it can be applied directly to lithography patterned substrates, meaning a high compatibility with the well-developed conventional top-down approaches to nanodevices. This bubble deposition technique is expected to be a promising method in the field of nano-object assembly and organization and has great application potentials.
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]yttrium
Sigma-Aldrich
Tris[N,N-bis(trimethylsilyl)amide]samarium(III), 98%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide, 97%
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1 M in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in tert-butyl methyl ether
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 40% in THF
Sigma-Aldrich
Hexamethyldisilazane, Wacker Chemie AG, ≥97.0% (GC)
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in hexanes
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.0 M in THF
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 0.5 M in toluene
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide solution, 0.6 M in toluene
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide solution, 1.0 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Sodium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 0.5 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1.5 M in THF
Supelco
Hexamethyldisilazane, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Potassium bis(trimethylsilyl)amide, 95%
Sigma-Aldrich
Lithium bis(trimethylsilyl)amide solution, 1 M in toluene