Merck
  • On-line chemiluminescence determination of mitoxantrone by capillary electrophoresis.
On-line chemiluminescence determination of mitoxantrone by capillary electrophoresis.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-09-08)
Suqin Han, Huili Wang
ABSTRACT
A novel capillary electrophoresis (CE) with chemiluminescence (CL) detection method for the determination of mitoxantrone (MTX) has been developed, which based on the CL reaction of potassium ferricyanide with luminol in sodium hydroxide medium sensitized by MTX. Under optimum analytical conditions, MTX is determined over the range of 7.0×10(-8)-1.0×10(-6)M with a detection limit of 1.0×10(-8)M. The relative standard deviation (RSD) was 3.7%, 2.6% and 3.0% for 7.0×10(-8), 5.0×10(-7) and 1.0×10(-6)M MTX (n=11), respectively. In laboratory-built CE-CL apparatus, the proposed method has been applied to determination of MTX in commercial drug and spiked in human urine and plasma with satisfactory results.
Sigma-Aldrich
Potassium ferricyanide(III), powder or chunks, <10 μm, 99%
Sigma-Aldrich
Potassium hexacyanoferrate(III), ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium hexacyanoferrate(III), BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Potassium hexacyanoferrate(III), 99.98% trace metals basis
Sigma-Aldrich
Potassium hexacyanoferrate(III), ReagentPlus®, ~99%