- Persistence of propargite on okra under subtropical conditions at Ludhiana, Punjab, India.
Persistence of propargite on okra under subtropical conditions at Ludhiana, Punjab, India.
Bulletin of environmental contamination and toxicology (2010-08-11)
B K Kang, Gagan Jyot, R K Sharma, R S Battu, B Singh
PMID20697883
ABSTRACT
Residues of propargite were estimated in okra fruits by high performance liquid chromatography following single application of Omite 57 EC @570 and 1,140 g a.i./ha. Residues of propargite were confirmed by high performance thin layer chromatography. The average initial deposits of propargite were observed to be 1.36 and 3.32 mg/kg, respectively, which were below its maximum residue limit of 2 mg/kg. The residue levels of propargite dissipated below its limit of quantification of 0.02 mg/kg after 7 days at single dose and after 10 days at double dose. The half-life values (T ₁(/)₂) of propargite were worked out to be 0.79 and 0.73 days, respectively, at recommended and double the recommended dosages.