Merck
  • Adsorption of p-nitroaniline from aqueous solutions onto activated carbon fiber prepared from cotton stalk.
Adsorption of p-nitroaniline from aqueous solutions onto activated carbon fiber prepared from cotton stalk.
Journal of hazardous materials (2009-01-23)
Kunquan Li, Zheng Zheng, Jingwei Feng, Jibiao Zhang, Xingzhang Luo, Guohua Zhao, Xingfa Huang
ABSTRACT
Activated carbon fiber prepared from cotton stalk was used as an adsorbent for the removal of p-nitroaniline (PNA) from aqueous solutions. Liquid phase adsorption experiments were conducted and the maximum adsorptive capacity was determined. The effect of experimental parameters such as pH, salinity and temperature on the adsorption was studied. The obtained experimental data were then fitted with the Langmuir, Freundlich and Redlich-Peterson models to describe the equilibrium isotherms. The kinetics rates were modeled by using the pseudo-first-order and pseudo-second-order equations. The results indicated that cotton stalk activated carbon fiber (CS-ACF) is an effective adsorbent for the removal of PNA from aqueous solutions. The maximum adsorption capacity of 406 mg g(-1) was achieved at the initial PNA concentration of 200 mg L(-1). The optimum pH for the removal of PNA was found to be 7.6. The presence of ammonium chloride proved to be favorable for the process of adsorption. The adsorption amount decreased with increasing temperature. The Redlich-Peterson model was found to best represent the equilibrium data. The kinetic data followed closely the pseudo-second-order equation. Thermodynamic study showed the adsorption was a spontaneous exothermic physical process.
Product Description
Supelco
4-Nitroaniline, analytical standard
Sigma-Aldrich
4-Nitroaniline, ≥99%