Merck
  • Identification of amino acids and domains required for catalytic activity of DPPR synthase, a cell wall biosynthetic enzyme of Mycobacterium tuberculosis.
Identification of amino acids and domains required for catalytic activity of DPPR synthase, a cell wall biosynthetic enzyme of Mycobacterium tuberculosis.
Microbiology (Reading, England) (2008-03-04)
Hairong Huang, Stefan Berg, John S Spencer, Danny Vereecke, Wim D'Haeze, Marcelle Holsters, Michael R McNeil
ABSTRACT
Decaprenylphosphoryl-d-arabinose (DPA) has been shown to be the donor of the essential d-arabinofuranosyl residues found in the cell wall of Mycobacterium tuberculosis. DPA is formed from phosphoribose diphosphate in a four-step process. The first step is the nucleophilic replacement of the diphosphate group with decaprenyl phosphate. This reaction is catalysed by the integral membrane protein 5-phospho-alpha-D-ribose-1-diphosphate : decaprenyl-phosphate 5-phosphoribosyltransferase (DPPR synthase). The enzyme is essential for growth and thereby an important target candidate for the development of new tuberculosis drugs. Although membrane proteins are an important subset of targets for current antibacterial agents, details about the structures and the active sites of such proteins are often not readily available by X-ray crystallography. To begin a different approach to the issue, homologues from Mycobacterium smegmatis and Corynebacterium glutamicum were expressed in Escherichia coli and shown to be active DPPR synthases. This was followed by bioinformatic analyses of the aligned sequences and then by site-directed mutagenesis of amino acids identified as likely to be important for activity. The results suggested that the enzymic synthesis of decaprenyl-phosphate 5-phosphoribose (DPPR) occurs on the cytoplasmic side of the plasma membrane. Amino acid substitutions showed that the predicted cytoplasmic N-terminal region and two cytoplasmic loops are involved in substrate binding and/or catalysis along with parts of some adjoining inner membrane regions. The enzyme lacks the classical phosphoribose diphosphate (pRpp) binding site found in nucleic acid precursor enzymes of both prokaryotes and eukaryotes but instead contains a conserved NDxxD motif required for enzymic activity. Thus, it is plausible that this DPPR synthase has a pRpp binding site that is different from that of the classical eukaryotic enzymes, and further work to develop inhibitors against this enzyme is thereby encouraged.
Sigma-Aldrich
D-Ribulose 1,5-bisphosphate sodium salt hydrate, ≥90% (TLC)
Sigma-Aldrich
D-Ribulose 1,5-bisphosphate sodium salt hydrate, ≥99.0% (TLC)