Merck
  • Effects of high-affinity inhibitors on partial reactions, charge movements, and conformational States of the Ca2+ transport ATPase (sarco-endoplasmic reticulum Ca2+ ATPase).
Effects of high-affinity inhibitors on partial reactions, charge movements, and conformational States of the Ca2+ transport ATPase (sarco-endoplasmic reticulum Ca2+ ATPase).
Molecular pharmacology (2008-01-24)
Francesco Tadini-Buoninsegni, Gianluca Bartolommei, Maria Rosa Moncelli, Daniel M Tal, David Lewis, Giuseppe Inesi
ABSTRACT
The inhibitory effects of thapsigargin, cyclopiazonic acid, and 2,5-di(tert-butyl)hydroquinone, and 1,3-dibromo-2,4,6-tri(methylisothiouronium)benzene on the Ca(2+) ATPase were characterized by comparative measurements of sequential reactions of the catalytic and transport cycle, including biochemical measurements and detection of charge movements within a single cycle. In addition, patterns of ATPase proteolytic digestion with proteinase K were derived to follow conformational changes through the cycle or after inhibitor binding. We find that thapsigargin, cyclopiazonic acid, and 2,5-di(tert-butyl)hydroquinone inhibit Ca(2+) binding and catalytic activation as demonstrated with isotopic tracers and lack of charge movement upon addition of Ca(2+) in the absence of ATP. It has been shown previously that binding of these inhibitors requires the E2 conformational state of the ATPase, obtained in the absence of Ca(2+). We demonstrate here that E2 state conformational features are in fact induced by these inhibitors on the ATPase even in the presence of Ca(2+). The resulting dead-end complex interferes with progress of the catalytic and transport cycle. Inhibition by 1,3-dibromo-2,4,6-tri(methylisothiouronium)benzene, on the other hand, is related to interference with a conformational transition of the phosphorylated intermediate (E1 approximately P . 2Ca(2+) to E2-P . 2Ca(2+) transition), as demonstrated by increased phosphoenzyme levels and absence of bound Ca(2+) translocation upon addition of ATP. This transition includes large movements of ATPase headpiece domains and transmembrane segments, produced through utilization of ATP-free energy as the "conformational work" of the pump. We conclude that the mechanism of high-affinity Ca(2+) ATPase inhibitors is based on global effects on protein conformation that interfere with ATPase cycling.
Sigma-Aldrich
2,5-Di-tert-butylhydroquinone, 99%