Merck
Comparison of composition and absorption of sugarcane policosanols.
The British journal of nutrition (2007-02-15)
Christopher P F Marinangeli, Amira N Kassis, Deepak Jain, Naoyuki Ebine, Stephen C Cunnane, Peter J H Jones
ABSTRACT
Policosanols (PC) exist as very-long-chain alcohols derived from sugarcane currently used in many countries as a cholesterol-lowering therapy. PC purity and relative percentage composition have been suggested as primary reasons why the original Cuban PC (OPC) supplements possess lipid-lowering efficacy. The purpose of the present study was, first, to compare the relative percentage purity and PC composition of both OPC and alternative sources of PC (APC). A second objective was to feed Syrian hamsters a diet containing 0.275 mg PC/g of either the OPC or an APC product (APC1) and compare subsequent tissue, plasma and faecal PC levels. Five animals from the APC1 dietary group received a diet containing ten times the original amount of PC. Results indicate that the APC formulations have a composition that is highly consistent with the OPC supplement, with octacosanol being present within the cited 60-70 % range. PC were undetectable in the small intestine, liver, adipose or plasma in animals fed either source. Hamsters fed OPC excreted octacosanol (C28) more rapidly (P < 0.05) than hamsters receiving APC1. If the cholesterol-lowering efficacy of PC mixtures is dependent on their purity and composition, then sugarcane-derived APC products should possess similar therapeutic properties as the OPC supplement.
Sigma-Aldrich
1-Octacosanol, ≥99% (GC)