Merck
  • Facile ring opening of 2,3-epoxy-steroids with aromatic amines in ionic liquids.
Facile ring opening of 2,3-epoxy-steroids with aromatic amines in ionic liquids.
Steroids (2006-06-06)
Anita Horváth, Rita Skoda-Földes, Sándor Mahó, Zoltán Berente, László Kollár
ABSTRACT
Efficient ring opening of steroidal 2,3-epoxides with stoichiometric amount of aromatic amines has been carried out using an ionic liquid ([bmim](+)[BF(4)](-)) both as solvent and catalyst. The reactions were completely regio- and stereoselective in each case. The aminoalcohol products have chair conformations in ring A. The ionic liquid-mediated ring opening can efficiently be carried out with aliphatic amines like morpholine as well.
Sigma-Aldrich
N-Methylaniline, purum, ≥98.0% (GC)
Supelco
N-Methylaniline, analytical standard
Sigma-Aldrich
N-Methylaniline, 98%
Sigma-Aldrich
N-Methylaniline, ≥99%