Merck
  • Performance of TX-100 and TX-114 for the separation of chrysoidine dye using cloud point extraction.
Performance of TX-100 and TX-114 for the separation of chrysoidine dye using cloud point extraction.
Journal of hazardous materials (2006-04-08)
M K Purkait, S DasGupta, S De
ABSTRACT
Cloud point extraction (CPE) is carried out to extract chrysoidine dye from aqueous solution using two different non-ionic surfactants, TX-100 and TX-114. The effects of different operating parameters, e.g., concentrations of surfactant, dye and salt, temperature, pH on extraction of both dye and surfactant have been studied in detail. The extraction of dye increases with temperature, surfactant concentration and salt concentration. Various design parameters of a CPE process have been estimated by developing correlations for dye solubilization and fractional coacervate phase volume with the operating conditions. The equilibrium solubilization data at four different temperatures follow Langmuir type isotherm. A method is presented to calculate the feed surfactant concentration required for the removal of dyes up to a level of 3.82x10(-6) M. The developed correlations may be useful to design a cloud point extractor of a desired efficiency.
Product Description
Supelco
Chrysoidine G, analytical standard
Sigma-Aldrich
Chrysoidine G