Merck
[Study of biocompatibility of polybutylcyanoacrylate nanoparticles].
Di 1 jun yi da xue xue bao = Academic journal of the first medical college of PLA (2005-10-20)
Xi-xiao Yang, Jan-hai Chen, Dan Guo
ABSTRACT
To evaluate the biocompatibility of polybutylcyanoacrylate nanoparticles (PBCA-NP) manufactured by emulsion polymerization technique. MTT assay was performed for evaluation of the cytotoxicity of PBCA-NP (e.g. cell proliferation and hemolytic test) experiments of long-term subcutaneous implantation and implantation in the muscular tissues of PBCA-NP was conducted in rabbits to observe the inflammatory reactions due to the implantation. No obvious cytotoxicity or hemolytic reactions were observed (with the hemolytic rate at different concentrations of PBCA-NP(5%). Three months after PBCA-NP implantation, obvious lymphocyte infiltration was not observed in the tissues around the implants which underwent gradual degradation. PBCA-NP possesses good biocompatibility.
Sigma-Aldrich
Polybutenes, average Mn ~920 by VPO, isobutylene >90 %
Sigma-Aldrich
Polybutenes, average Mn ~2,300 by VPO, isobutylene >90 %