Merck
Surfactant-free synthesis and functionalization of gold nanoparticles.
Journal of the American Chemical Society (2005-09-15)
Martin Schulz-Dobrick, K Vijaya Sarathy, Martin Jansen
ABSTRACT
A new, facile and generally applicable synthesis of functionalized gold nanoparticles is presented. It is based on the surfactant-free generation of weakly stabilized nanoparticles by the reduction of HAuCl4 with sodium naphthalenide in diglyme. These nanoparticles were found to lack long-term stability. However, stabilization in both unpolar and polar solvents could straightforwardly be achieved by subsequent addition of various capping ligands. The resulting ligand-capped gold nanoparticles were investigated by TEM microscopy, UV-vis, and FT-IR spectroscopy. Particle core size can be tuned by the amount of reduction agent. The strict separation of the reduction step and the functionalization step in this one-pot synthesis offers an easy and fast access to highly functionalized gold nanoparticles.
Product Description
Supelco
Diethylene glycol dimethyl ether, analytical standard
Sigma-Aldrich
Diethylene glycol dimethyl ether, anhydrous, 99.5%