Merck
  • Palladium-catalyzed annulation of arynes by 2-halobenzaldehydes: synthesis of fluoren-9-ones.
Palladium-catalyzed annulation of arynes by 2-halobenzaldehydes: synthesis of fluoren-9-ones.
Organic letters (2005-08-27)
Xiaoxia Zhang, Richard C Larock
ABSTRACT
Arynes, generated in situ from 2-(trimethylsilyl)aryl triflates and CsF, undergo annulation by o-haloarenecarboxaldehydes in the presence of a Pd catalyst, providing a useful new method for the synthesis of fluoren-9-ones in good yields. [reaction: see text]
Sigma-Aldrich
2-Bromobenzaldehyde, 98%
Sigma-Aldrich
9-Fluorenone, 98%