Merck
  • GC/MS identification and quantification of constituents of bacterial lipids and glycoconjugates obtained after methanolysis as heptafluorobutyrate derivatives.
GC/MS identification and quantification of constituents of bacterial lipids and glycoconjugates obtained after methanolysis as heptafluorobutyrate derivatives.
Analytical biochemistry (2005-04-21)
Anne Bohin, Franck Bouchart, Colette Richet, Osarath Kol, Yves Leroy, Philippe Timmerman, Guillemette Huet, Jean-Pierre Bohin, Jean-Pierre Zanetta
ABSTRACT
In previous articles [Anal. Biochem. 284 (2000) 201; J. Lipid Res. 43 (2002) 794], we reported that the GC/MS identification and quantification of nearly all constituents of glycolipids could be obtained on the same sample in a single GC/MS analysis as heptafluorobutyrate derivatives of the products liberated using acid-catalyzed methanolysis. The same type of data could be obtained on glycoproteins and proteoglycans [Biochemistry 42 (2003) 8342]. These experiments were performed on material from higher organisms, and there was no evidence that bacteria-specific constituents could also be identified and quantified. The current article reports that the GC/MS analysis of compounds liberated by acid-catalyzed methanolysis as heptafluorobutyrate derivatives allows the simultaneous qualitative and quantitative determinations of pentoses, deoxyhexoses, hexoses, hexosamines, uronic acids, Kdo, Mur, heptose, Kdn, and neuraminic acid as well as of most fatty acids (including hydroxylated fatty acids). This approach provides a way of obtaining fingerprints of bacterial constituents and quantification of the overall effect of gene inactivation or of culture conditions.
Sigma-Aldrich
Heptafluorobutyric anhydride, ≥98%
Supelco
Heptafluorobutyric anhydride, LiChropur, ≥99.0%
Supelco
Heptafluorobutyric anhydride, derivatization grade (GC derivatization), LiChropur, ≥99.0%