Merck
  • An Au-catalyzed cyclialkylation of electron-rich arenes with epoxides to prepare 3-chromanols.
An Au-catalyzed cyclialkylation of electron-rich arenes with epoxides to prepare 3-chromanols.
Journal of the American Chemical Society (2004-05-13)
Zhangjie Shi, Chuan He
ABSTRACT
A gold-catalyzed cyclialkylation of electron-rich arenes with tethered epoxides afforded 3-chromanols stereospecifically.
Sigma-Aldrich
1,2,3-Trimethoxybenzene, 98%