Merck
  • Choline pivaloyl esters improve in rats cognitive and memory performances impaired by scopolamine treatment or lesions of the nucleus basalis of Meynert.
Choline pivaloyl esters improve in rats cognitive and memory performances impaired by scopolamine treatment or lesions of the nucleus basalis of Meynert.
Neuroscience letters (2004-03-24)
Vincenzo Rispoli, Domenicantonio Rotiroti, Vincenzo Carelli, Felice Liberatore, Luigi Scipione, Rosario Marra, Gianfabio Giorgioni, Antonio Di Stefano
ABSTRACT
The effects of two choline pivaloyl esters, [2-(2,2-dimethylpropionyloxy)ethyl]trimethylammonium iodide (1) and [2-(2,2-dimethylpropionyloxy)ethyl]trimethylammonium 2,2-dimethylpropionate (2), on learning and memory impairments induced in rats by scopolamine or lesions of nucleus basalis magnocellularis (NBM) have been evaluated by object recognition and Morris water maze tests in comparison with Tacrine (THA). Both 1 and 2 restored discrimination in object recognition test for assessing working-episodic memory and improved spatial memory in scopolamine or NBM-lesioned rats as well. The positive effects produced by 1 and 2 on cognitive and memory deficits were well comparable with those evoked by THA, used as reference compound.
Sigma-Aldrich
Trimethylphenylammonium chloride, ≥98%
Sigma-Aldrich
Trimethylphenylammonium bromide, 98%
Sigma-Aldrich
Trimethylphenylammonium hydroxide solution, ~25% in H2O (1.68 M)
Supelco
Trimethylphenylammonium hydroxide solution, ~0.5 M (CH3)3N(OH)C6H5 in methanol, for GC derivatization, LiChropur
Sigma-Aldrich
Trimethylphenylammonium tribromide, 97%