Merck
  • Time-dependent mechanical properties of polymeric coatings used in rupturable pulsatile release dosage forms.
Time-dependent mechanical properties of polymeric coatings used in rupturable pulsatile release dosage forms.
Drug development and industrial pharmacy (2003-08-02)
T Bussemer, N A Peppas, R Bodmeier
ABSTRACT
The mechanical properties of polymer films used in pharmaceutical coatings of pulsatile drug delivery systems were evaluated in the dry and the wet state by a newly developed puncture test, which allowed the time-dependent measurement of the mechanical properties on the same film specimen. Force, puncture strength, energy at break, modulus, and strain were investigated as a function of water exposure time with respect to the type of polymer and the type and concentration of plasticizer and pore former (hydroxypropyl methylcellulose, HPMC). Eudragit RS films were very flexible, had a high strain, and broke upon puncture with only small cracks. In contrast, ethylcellulose films were more brittle with a lower strain and showed complete film rupture. Increased amounts of the hydrophilic pore former, HPMC, resulted in a reduced puncture strength and in an increase in water uptake and weight loss of the films. The puncture strength decreased with increasing plasticizer concentration and was lower with the lipophilic dibutyl sebacate than with the hydrophilic triethyl citrate.
Sigma-Aldrich
Dibutyl sebacate, technical grade
Supelco
Dibutyl sebacate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dibutyl sebacate, ≥97.0% (GC)