Merck
  • Crystallization and preliminary X-ray diffraction studies of the transcriptional regulator TraR bound to its cofactor and to a specific DNA sequence.
Crystallization and preliminary X-ray diffraction studies of the transcriptional regulator TraR bound to its cofactor and to a specific DNA sequence.
Acta crystallographica. Section D, Biological crystallography (2002-07-24)
Alessandro Vannini, Cinzia Volpari, Cesare Gargioli, Ester Muraglia, Raffaele De Francesco, Petra Neddermann, Stefania Di Marco
ABSTRACT
TraR is an Agrobacterium tumefaciens transcriptional regulator which binds the pheromone N-3-oxooctanoyl-L-homoserine lactone (AAI) in response to the bacterial population density. The TraR-AAI complex dimerizes and interacts with a specific 18-base-pair DNA sequence (TraBox), activating promoters containing this site. TraR was overexpressed and purified from Escherichia coli. Crystals of the ternary complex, in which dimeric TraR-AAI is bound to the TraBox sequence, have been obtained by the vapour-diffusion method. The crystals belong to space group P2(1)2(1)2(1), with unit-cell parameters a = 66.99, b = 94.67, c = 209.66 A, with two (TraR-AAI)(2)-TraBox complexes in the asymmetric unit. A three-wavelength MAD data set for the seleno-L-methionine-substituted form has been collected to a resolution of 3 A. 20 of the 24 crystallographically independent selenium sites were located as part of the MAD-phasing process.
Sigma-Aldrich
N-(3-Oxooctanoyl)-L-homoserine lactone, ≥97% (HPLC), white, powder