Merck
  • Synthesis of erythro and threo furanoid glycals from 1- and 2-phenylselenenyl-carbohydrate derivatives.
Synthesis of erythro and threo furanoid glycals from 1- and 2-phenylselenenyl-carbohydrate derivatives.
Carbohydrate research (2001-11-02)
F Bravo, M Kassou, Y Díaz, S Castillón
ABSTRACT
Differently protected erythro and threo furanoid glycals were synthesized by selenoxide elimination when phenyl 1-selenoglycosides were treated in oxidizing conditions (tBuOOH, Ti(O(i)Pr)(4), Et(2)(i)PrN). The phenyl 1-selenoglycosides were obtained from methyl 2-deoxy-D-erythro-pentofuranoside by protection of the primary hydroxyl or both hydroxyls and further reaction with PhSeH in the presence of BF(3).Et(2)O. Erythro and threo furanoid glycals were also prepared by treating 2-deoxy-2-phenylselenenyl-1,4-anhydrocyclitols under similar conditions. The 2-deoxy-2-phenylselenenyl-1,4-anhydrocyclitols were obtained from 4-pentene-1,2,3-triols by a 5-endo selenium electrophilic induced cyclization.
Sigma-Aldrich
D-Threitol, 99%