Merck
  • Chiral separation of homocysteine by derivatization with 4-aminosulfonyl-7-fluoro-2,1,3-benzoxadiazole followed by capillary electrophoresis using gamma-cyclodextrin.
Chiral separation of homocysteine by derivatization with 4-aminosulfonyl-7-fluoro-2,1,3-benzoxadiazole followed by capillary electrophoresis using gamma-cyclodextrin.
Journal of chromatography. A (2000-06-14)
I J Kim, S J Park, H J Kim
ABSTRACT
Homocysteine was derivatized with 4-aminosulfonyl-7-fluoro-2,1,3-benzoxadiazole (ABD-F) to form an inclusion complex with cyclodextrin and to facilitate UV detection. ABD-homocysteine showed interaction with beta- and gamma-cyclodextrin in capillary electrophoresis at pH 2.25 as indicated by the decreased migration time. However, chiral separation of D,L-ABD-homocysteine was observed using gamma-CD only. Optimal separation was obtained at pH 2.25, 50 mM gamma-CD concentration, and 20 kV applied voltage. L-ABD-Homocysteine migrated faster than the D-isomer as demonstrated by a spiking experiment using dithiothreitol-reduced L-homocystine.
Sigma-Aldrich
4-Fluoro-7-sulfamoylbenzofurazan