Merck
Metabolism of [(14)C]flusilazole in the goat.
Journal of agricultural and food chemistry (2000-05-04)
J J Anderson, L M Shalaby, D S Berg
ABSTRACT
[Phenyl(U)-(14)C] and [triazole(3)-(14)C]flusilazole ([(bis 4-fluorophenyl)]methyl(1H-1,2,4-triazole-1-ylmethyl)silane; I) were extensively metabolized when fed to lactating goats (Capra hircus). The primary metabolites identified in goat tissues and milk were bis(4-fluorophenyl)(methyl)silanol (II) and 1H-1,2,4-triazole (III). Concentrations of total radiolabeled residues in the milk ranged from 0.09 to 0.74 microg/mL. Concentrations of radiolabeled residues found in tissues when the [(14)C] label was in the phenyl or triazole position, respectively, were 13.5 and 3.54 microg/g (liver), 8.74 and 0.75 microg/g (kidney), 0.41 and 0.52 microg/g (leg muscle), and 4.07 and 0.94 microg/g (back fat). Urine contained an additional major metabolite identified as [bis(4-fluorophenyl)](methyl)silylmethanol (IV) and its glucuronic acid conjugate (V). With either labeled form of flusilazole, the majority of the recovered radiolabel was excreted in urine or feces.
Product Description
Supelco
Flusilazole, PESTANAL®, analytical standard