- Spectrophotometry assay of imipramine and desipramine using ammonium metavanadate and its application to pharmaceutical preparations.
Spectrophotometry assay of imipramine and desipramine using ammonium metavanadate and its application to pharmaceutical preparations.
Journal of pharmaceutical and biomedical analysis (2000-03-22)
W Misiuk
PMID10727139
ABSTRACT
Simple and sensitive method for determination of imipramine and desipramine is reported. The procedure is based on the oxidation of the drugs by ammonium metavanadate. Linear calibration graphs were obtained in the concentration range 0.6-40 microg ml(-1) of imipramine and 0.7-35 microg ml(-1) of desipramine with a relative standard deviation (RSD) less than 0.5%. The method was applied to the determination of the drugs in pharmaceutical preparations and compared favourably with independent official methods.