Merck
  • Endogenous secretory RAGE in obese women: association with platelet activation and oxidative stress.
Endogenous secretory RAGE in obese women: association with platelet activation and oxidative stress.
The Journal of clinical endocrinology and metabolism (2012-07-05)
Natale Vazzana, Maria Teresa Guagnano, Chiara Cuccurullo, Elisabetta Ferrante, Stefano Lattanzio, Rossella Liani, Mario Romano, Giovanni Davì
ABSTRACT
The receptor for advanced glycation end-products (RAGE) has been implicated in obesity-related metabolic disease and accelerated atherothrombosis. We tested the hypothesis that changes in endogenous secretory (es)RAGE levels as a result of excess adiposity and oxidative stress may contribute to enhancing platelet activation in obese women, thus increasing the cardiovascular risk. Eighty otherwise healthy obese women and 20 nonobese women were studied. esRAGE and plasma adiponectin were reduced in obese women [median (interquartile range), 0.18 (0.13-0.26) vs. 0.38 (0.20-0.48) ng/ml, P = 0.003; and 4.4 (2.8-6.4) vs. 10.0 (6.9-12.5) μg/ml, P < 0.0001, respectively] who also displayed higher urinary 11-dehydro-thromboxane B(2) (11-dehydro-TXB(2)) [795 (572-1089) vs. 211 (135-301) pg/mg creatinine; P < 0.0001] and 8-iso-prostaglandin F(2α) (8-iso-PGF(2α)) [544 (402-698) vs. 149 (98-219) pg/mg creatinine; P < 0.0001] compared to nonobese women. Direct correlations between plasma adiponectin and esRAGE (Rho = 0.43; P < 0.0001) and between urinary 8-iso-PGF(2α) and 11-dehydro-TXB(2) (Rho = 0.36; P = 0.001) were observed in obese women. Moreover, plasma esRAGE and urinary 11-dehdro-TXB(2) were inversely related (Rho = -0.29; P = 0.008). On multiple linear regression analysis, urinary 8-iso-PGF(2α) and plasma esRAGE were independent predictors of urinary 11-dehydro-TXB(2). In five obese women, a short-term weight loss program gave a significant increase in esRAGE and decrease in urinary 8-iso-PGF(2α) and 11-dehydro-TXB(2). In otherwise healthy obese women, low plasma esRAGE levels are associated with reduced circulating adiponectin and enhanced thromboxane biosynthesis, which is in part mediated by increased lipid peroxidation. Thus, excess adiposity may be implicated in RAGE hyperactivation and thromboxane-dependent platelet activation, contributing to obesity-related metabolic and vascular disease.