Merck
  • Sarafotoxin S6c: an agonist which distinguishes between endothelin receptor subtypes.
Sarafotoxin S6c: an agonist which distinguishes between endothelin receptor subtypes.
Biochemical and biophysical research communications (1991-03-25)
D L Williams, K L Jones, D J Pettibone, E V Lis, B V Clineschmidt
ABSTRACT
In contrast to endothelin-1 (ET-1) and several of its analogues, sarafotoxin S6c (S6c) was a much more potent inhibitor of [125I]-ET-1 binding in rat hippocampus and cerebellum (Ki approximately 20 pM) than in rat atria and aorta (Ki approximately 4500 nM), suggesting the existence of ET-1 receptor subtypes (aorta/atria, ETA; hippocampus/cerebellum, ETB). S6c was a potent activator of PI turnover in hippocampus (EC50 approximately 10 nM) but not atria (EC50 greater than 1 microM), unlike ET-1 which was active in both tissues. S6c, therefore, is a highly selective ETB agonist. Furthermore, S6c was a potent pressor agent in the pithed rat (ED25 mm Hg approximately 0.1 nmoles/kg, i.v.), suggesting that the ETB receptor subtype may be important in cardiovascular function.
Sigma-Aldrich
Sarafotoxin S6c, ≥97% (HPLC)