Merck
  • Bile acid modulates transepithelial permeability via the generation of reactive oxygen species in the Caco-2 cell line.
Bile acid modulates transepithelial permeability via the generation of reactive oxygen species in the Caco-2 cell line.
Free radical biology & medicine (2005-08-20)
Yoshio Araki, Takuji Katoh, Atsushi Ogawa, Shigeki Bamba, Akira Andoh, Shigeki Koyama, Yoshihide Fujiyama, Tadao Bamba
ABSTRACT
The barrier functions in epithelial and endothelial cells seem to be very important for maintaining normal biological homeostasis. However, it is unclear whether or how bile acids affect the epithelial barrier. We examined the bile acid-induced disruption of the epithelial barrier. We measured the transepithelial electrical resistance (TEER) of Caco-2 cells as a marker of disruption of the epithelial barrier. Reactive oxygen species (ROS) generation was also measured. Cholic acid (CA) decreased the TEER and increased intracellular ROS generation. PLA2 (phospholipase A2), COX (cyclooxygenase), PKC (protein kinase), ERK 1/2 (extracellular signal-regulated kinase 1/2), PI 3 K (phosphatidylinositol 3-kinase), p38 MAPK (p38 mitogen-activated protein kinase), MLCK (myosin light-chain kinase), NADH dehydrogenase, and XO (xanthine oxidase) inhibitors or ROS scavengers prevented the CA-induced TEER decrease. PLA2, COX, PKC, NADH dehydrogenase, and XO inhibitors prevented the CA-induced ROS generation but not ERK 1/2, PI 3 K, p38 MAPK, and MLCK inhibitors. If the cells were treated with ROS generators such as superoxide dismutase, the TEER decreased. ERK 1/2, PI 3 K, p38 MAPK, and MLCK inhibitors prevent these ROS generators from inducing the TEER decrease. These results suggest that ROS play an important role. In addition, PLA2, COX, PKC, NADH dehydrogenase, and XO are located upstream of the ROS generation, but ERK 1/2, PI 3 K, p38 MAPK, and MLCK are downstream during the signaling of CA-induced TEER alterations.
Sigma-Aldrich
OBAA, ≥98% (HPLC)